EMAIL THIS PAGE TO A FRIEND
Pharmaceutical research
Relationship between particle size and impurity incorporation during crystallization of (+)-pseudoephedrine hydrochloride, acetaminophen, and adipic acid from aqueous solution.
PMID 12180541
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
456071
Calcium sulfate, with indicator, 6 mesh
CaO4S