EMAIL THIS PAGE TO A FRIEND
Cellular and molecular life sciences : CMLS
RAGE is a multiligand receptor of the immunoglobulin superfamily: implications for homeostasis and chronic disease.
PMID 12222959
Abstract
Receptor for AGE (RAGE) is a member of the immunoglobulin superfamily that engages distinct classes of ligands. The biology of RAGE is driven by the settings in which these ligands accumulate, such as diabetes, inflammation, neurodegenerative disorders and tumors. In this review, we discuss the context of each of these classes of ligands, including advance glycation end-products, amyloid beta peptide and the family of beta sheet fibrils, S100/calgranulins and amphoterin. Implications for the role of these ligands interacting with RAGE in homeostasis and disease will be considered.