EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Alternative splicing products of the gene for a human nuclear actin-related protein, hArpNbeta/Baf53, that encode a protein isoform, hArpNbetaS, in the cytoplasm.
PMID 12353638
Abstract
A human nuclear actin-related protein, hArpNbeta/ Baf53, is a component of chromatin remodeling and histone acetyltransferase complexes. We identified two alternative splicing products of the gene for hArpNbeta/ Baf53. They encoded a protein isoform, hArpNbetaS; and its fusion product with green fluorescent protein was to be found in the cytoplasm, not the nucleus. The isoforms may contribute to functional regulation of these complexes.
Related Materials