EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Novel synthesis of 2-chloroquinolines from 2-vinylanilines in nitrile solvent.
PMID 12398521
Abstract
2-Vinyl- or heteroaryl-substituted anilines were reacted with diphosgene in acetonitrile solution via a reactive imidoyl moiety to afford the corresponding 2-chloroquinolines. Facile syntheses of nine 2-chloroquinoline derivatives from several anilines and their postulate mechanism is described. The postulate mechanism of 2-chloroquinoline formation via imidoyl moiety as a good leaving group shows that the reaction consists of the following three steps: (1) generation of phenylisocyanate, (2) quinoline ring formation, and (3) chlorination on C2 position of quinoline.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
557803
2,6-Dichloroquinoline, 97%
C9H5Cl2N