EMAIL THIS PAGE TO A FRIEND
Diabetes
Induction of autoimmune diabetes through insulin (but not GAD65) DNA vaccination in nonobese diabetic and in RIP-B7.1 mice.
PMID 12401715
Abstract
Insulin has been used to modify T-cell autoimmunity in experimental models of type 1 diabetes. In a large clinical trial, the effect of insulin to prevent type 1 diabetes is currently investigated. We here show that insulin can adversely trigger autoimmune diabetes in two mouse models of type 1 diabetes, using intramuscular DNA vaccination for antigen administration. In female nonobese diabetic (NOD) mice, diabetes development was enhanced after preproinsulin (ppIns) DNA treatment, and natural diabetes resistance in male NOD mice was diminished by ppIns DNA vaccination. In contrast, GAD65 DNA conferred partial diabetes protection, and empty DNA plasmid was without effect. In RIP-B7.1 C57BL/6 mice (expressing the T-cell costimulatory molecule B7.1 in pancreatic beta-cells), autoimmune diabetes occurred in 70% of animals after ppIns vaccination, whereas diabetes did not develop spontaneously in RIP-B7.1 mice or after GAD65 or control DNA treatment. Diabetes was characterized by diffuse CD4(+)CD8(+) T-cell infiltration of pancreatic islets and severe insulin deficiency, and ppIns, proinsulin, and insulin DNA were equally effective for disease induction. Our work provides a new model of experimental autoimmune diabetes suitable to study mechanisms and outcomes of insulin-specific T-cell reactivity. In antigen-based prevention of type 1 diabetes, diabetes acceleration should be considered as a potential adverse result.