EMAIL THIS PAGE TO A FRIEND
Cytogenetic and genome research
Assignment of tumor protein p53 induced nuclear protein 1 (TP53INP1) gene to human chromosome band 8q22 by in situ hybridization.
PMID 12438758