EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Butyrate down-regulates CD44 transcription and liver colonisation in a highly metastatic human colon carcinoma cell line.
PMID 12439723
Abstract
Over-expression of the adhesion molecule CD44 and its splice variants, especially CD44v6, is associated with poor prognosis and metastasis. We aimed at regulating the expression of CD44 in the highly metastatic human colon cancer cell line HM7 and thereby affecting its metastatic ability. HM7 cells show constitutive expression of CD44 standard and variants isoforms, which were significantly down-regulated by treatment with butyrate. Butyrate significantly inhibited transcription of the CD44 gene and abolished epidermal growth factor-mediated up-regulation of the reporter gene luciferase subcloned upstream to the CD44 promoter (-1.1 kb) and transfected to HM7 cells. Nuclear proteins from butyrate-treated cells bound to an epidermal growth factor receptor element motif present in the CD44 promoter. Epidermal growth factor receptor element-site directed mutations eliminated the inducibility of the luciferase reporter gene and did not allowed binding of nuclear proteins harvested from butyrate-treated cells. Butyrate induced CD44 gene repression by specifically interacting with an epidermal growth factor receptor element nuclear transcriptional factor. This interaction affects CD44 transcriptional activity vis-à-vis in vivo metastatic ability of HM7 cells. These results provide additional insight into the anticarcinogenic properties of butyrate.