EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Cutting edge: carbohydrate profiling identifies new pathogens that interact with dendritic cell-specific ICAM-3-grabbing nonintegrin on dendritic cells.
PMID 12574325
Abstract
Dendritic cells (DC) are instrumental in handling pathogens for processing and presentation to T cells, thus eliciting an appropriate immune response. C-type lectins expressed by DC function as pathogen-recognition receptors; yet their specificity for carbohydrate structures on pathogens is not fully understood. In this study, we analyzed the carbohydrate specificity of DC-specific ICAM-3-grabbing nonintegrin (SIGN)/CD209, the recently documented HIV-1 receptor on DC. Our studies show that DC-SIGN binds with high affinity to both synthetic mannose- and fucose-containing glycoconjugates. These carbohydrate structures are abundantly expressed by pathogens as demonstrated by the affinity of DC-SIGN for natural surface glycans of the human pathogens Mycobacterium tuberculosis, Helicobacter pylori, Leishmania mexicana, and Schistosoma mansoni. This analysis expands our knowledge on the carbohydrate and pathogen-specificity of DC-SIGN and identifies this lectin to be central in pathogen-DC interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44367
Lewis-a trisaccharide, ≥98.0% (TLC)
C20H35NO15