EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Pd(OH)2/C (Pearlman's catalyst): a highly active catalyst for Fukuyama, Sonogashira, and Suzuki coupling reactions.
PMID 12585906
Abstract
Treatment of thiol esters 1 with zinc reagent 2 in the presence of a small amount (
Related Materials