EMAIL THIS PAGE TO A FRIEND
In vitro
Human colonic adenocarcinoma cells. I. Establishment and description of a new line.
PMID 1262041
Abstract
A series of human colonic epithelial cell lines have been cultured from a single patient: LS-180 the original adenocarcinoma, LS-174T a trypsinized variant, and normal colonic tissue. The malignant cells, 20 to 40, mum in diameter and oval to polygonal, exhibited characteristics of normal colonic mucosal cells, namely, abundant microvilli prominent in secretory cells, and the presence of intracytoplasmic mucin vacuoles. The cultured adenocarcinoma cells, but not normal, demonstrated neoplastic properties by producing high levels of carcinoembryonic antigen (CEA) and by the ability to be propagated in hamster cheek pouches and in immunodeprived mice. The CEA production by the newly established line LS-180 released 900 times more CEA per cell into the culture medium and bore 30 times more cell-associated material than the established line, HT-29. These cell lines may permit detection of distinctive chemical, physiological, pharmacologic, and immunologic characteristics of neoplastic colonic cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87060401
LS174T
87021202
LS180