EMAIL THIS PAGE TO A FRIEND
Nature
Transformation of C3H/10T1/2CL8 mouse embryo fibroblasts by ultraviolet irradiation and a phorbol ester.
PMID 1264242
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90110523
C3H/MCA clone 16