EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis of potent oxindole CDK2 inhibitors.
PMID 12659774
Abstract
A series of oxindole CDK2 inhibitors was synthesized. These novel analogues have a saturated monosubstituted cyclic moiety at their C-4 position that mimics the ribofuranoside of ATP. This substitution afforded agents with increased potency relative to the parent indolinone and nanomolar range IC(50) against the CDK2 enzyme and two cancer cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
540935
4-(N-Boc-amino)piperidine, 96%
C10H20N2O2