EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Linking the T cell surface protein CD2 to the actin-capping protein CAPZ via CMS and CIN85.
PMID 12690097
Abstract
Recruitment of CD2 to the immunological synapse in response to antigen is dependent on its proline-rich cytoplasmic tail. A peptide from this region (CD2:322-339) isolated CMS (human CD2AP); a related protein, CIN85; and the actin capping protein, CAPZ from a T cell line. In BIAcore analyses, the N-terminal SH3 domains of CMS and CIN85 bound CD2:322-339 with similar dissociation constants (KD = approximately 100 microm). CAPZ bound the C-terminal half of CMS and CIN85. Direct binding between CMS/CIN85 and CAPZ provides a link with the actin cytoskeleton. Overexpression of a fragment from the C-terminal half or the N-terminal SH3 domain of CD2AP in a mouse T cell hybridoma resulted in enhanced interleukin-2 production and reduced T cell receptor down-modulation in response to antigen. These adaptor proteins are important in T cell signaling consistent with a role for CD2 in regulating pathways initiated by CMS/CIN85 and CAPZ.