EMAIL THIS PAGE TO A FRIEND
Comprehensive therapy
The antidepressant sertraline: a review of its uses in a range of psychiatric and medical conditions.
PMID 12701343
Abstract
Although sertraline was initially introduced as an antidepressant, it can be prescribed for a wide range of psychiatric and medical conditions. We review the pharmacology, the adverse effects, the dosing guidelines, as well as the indications of sertraline.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6319
Sertraline hydrochloride, ≥98% (HPLC)
C17H17NCl2 · HCl