EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Efficient and practical method for synthesizing optically active indan-2-ols by the Ti(O-i-Pr)(4)/2 i-PrMgCl-mediated metalative Reppe reaction.
PMID 12790616
Abstract
An efficient and practical synthesis of optically active indan-2-ols 1 has been developed starting from readily accessible optically active 4-siloxy-1,6-alkadiynes 2 and ethynyl p-tolyl sulfone, where the metalative Reppe reaction mediated by an economical divalent titanium reagent, Ti(O-i-Pr)(4)/2 i-PrMgCl, is a key step.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
419869
Ethynyl p-tolyl sulfone, 98%
C9H8O2S