EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Forced expression of antisense 14-3-3beta RNA suppresses tumor cell growth in vitro and in vivo.
PMID 12896901
Abstract
The 14-3-3 family proteins are key regulators of various signal transduction pathways including malignant transformation. Previously, we found that the expression of the 14-3-3beta gene is deregulated as well as c-myc gene in aflatoxin B1 (AFB1)-induced rat hepatoma K1 and K2 cells. To elucidate the implication of 14-3-3beta in tumor cell growth, in this paper we analyzed the effect of forced expression of antisense 14-3-3beta RNA on the growth and tumorigenicity of K2 cells. K2 cells transfected with antisense 14-3-3beta cDNA expression vector diminished their growth ability in monolayer culture and in semi-solid medium. Expression level of vascular endothelial growth factor mRNA was also reduced in these transfectants. Tumors that formed by the transfectants in nude mice were much smaller and histologically more benign tumors, because of their decreased level of mitosis compared with those of the parental cells. Frequency of apoptosis detected by terminal deoxynucleotidyl transferase-mediated dUTP nick end labeling assay was increased in the transfectant-derived tumors accompanying the inhibition of angiogenesis. In addition, over-expression of 14-3-3beta mRNA was observed in various murine tumor cell lines. These results suggest that 14-3-3beta gene plays a pivotal role in abnormal growth of tumor cells in vitro and in vivo.