EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Activation of the ERK and JNK signaling pathways caused by neuron-specific inhibition of PP2A in transgenic mice.
PMID 12937125
Abstract
A reduced activity of protein phosphatase 2A (PP2A) has been shown in brains of patients with Alzheimer's disease (AD), a neurodegenerative disorder characterized histopathologically by amyloid plaques and neurofibrillary tangles. Tau, as the principal component of neurofibrillary tangles, can be hyperphosphorylated by a reduced activity of PP2A in vitro and by pharmacological approaches, suggesting a crucial role of PP2A in tangle formation. To dissect the role of PP2A in vivo, we previously generated transgenic mice with chronically reduced PP2A activity by expressing a dominant-negative mutant form of the PP2A catalytic subunit Calpha, L199P, under the control of a neuron-specific promoter. In these mice, endogenous tau is phosphorylated at the epitopes Ser202/Thr205 and Ser422. In vitro, these tau phospho-epitopes can be phosphorylated by the kinases ERK and JNK, and the kinases themselves are negatively regulated by PP2A. In this study, we show that chronic inhibition of PP2A activity in L199P transgenic mice causes the activation of ERK and JNK as demonstrated by the phosphorylation and nuclear accumulation of the ERK and JNK substrates, Elk-1 and c-Jun. TUNEL staining revealed that activated JNK signaling was not associated with cell death. Our findings imply that PP2A is a negative regulator of the ERK and JNK signaling pathways in vivo, suggesting that in AD, tau hyperphosphorylation may be caused in part by PP2A dysfunction.