EMAIL THIS PAGE TO A FRIEND
Protein science : a publication of the Protein Society
The synthetic peptide Gly-Pro-Arg-Pro-amide limits the plasmic digestion of fibrinogen in the same fashion as calcium ion.
PMID 1304901
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1895
Gly-Pro-Arg-Pro, ≥97% (HPLC)
C18H31N7O5
G5779
Gly-Pro-Arg-Pro amide, ≥97% (HPLC)
C18H32N8O4