EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Studies on the cyclic 3':5'-AMP-stimulated pig liver protein kinase reaction with pyruvate kinase as substrate.
PMID 13074
Abstract
The phosphorylation of pig liver pyruvate kinase by cyclic adenosine 3':5'-monophosphate-dependent protein kinase has been studied. For comparison, mixed histone and a synthetic heptapeptide were also used as substrates. Protein kinase was purified by chromatography on DEAE-cellulose, hydroxyapatite, and Sephadex G-200. The enzyme was stimulated by cyclic AMP with apparent Ka values of 2.5 and 0.8 x 10-7 M for pyruvate kinase and histone substrates, respectively. Divalent cations were essential for the activity of the protein kinase. Variation of the concentration of ATP resulted in approximately straight lines in Lineweaver-Burk plots for the phosphorylation of both pyruvate kinase and mixed histone. The apparent Km values for ATP were 21 and 11 muM, respectively. The phosphorylation rate increased with the concentration of pyruvate kinase even at a concentration of 2 muM pyruvate kinase. At a high ionic strength, the phosphorylation rate of both pyruvate kinase and histone decreased. The phosphorylation rate varied markedly with pH in imidazole/HC1 and Tris/HC1 buffers. At slightly alkaline pH values, pyruvate kinase was phosphorylated at a much higher rate than pH7, but this was not the case for histone. At pH 8.5, the phosphorylation rate of pyruvate kinase was 3.5 times the rate at pH 7, while the corresponding increase for the histone phosphorylation was 50 per cent. In potassium phosphate buffers, the phosphorylation rate of both substrates did not change significantly over the pH range studied. Arrhenius' plots of the protein kinase reaction resulted in a break at about 10 degrees when pyruvate kinase was used as substrate, whereas a straight line was obtained when using histone. The negative allosteric effectors of pyruvate kinase, alanine, and phenylalanine, increased the phosphorylation rate of pyruvate kinase at pH 8 by 50 and 120 per cent, respectively. The same effectors did not influence the phosphorylation rate of mixed histone or a synthetic heptapeptide. It is concluded that the conformations adopted by pyruvate kinase in the presence of allosteric inhibitors make it a better substrate for the protein kinase.