EMAIL THIS PAGE TO A FRIEND
Cancer research
Transplantable human neoplasms maintained in cortisone-treated laboratory animals: H.S. No. 1; H.Ep. No. 1; H.Ep. No. 2; H.Ep. No. 3; and H.Emb.Rh. No. 1.
PMID 13209540