EMAIL THIS PAGE TO A FRIEND
Annales de l'Institut Pasteur
[A selective medium for isolation of Brucella melitensis].
PMID 13218364
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3051 Brucella broth base, for microbiology