EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Affinity chromatographic purification of bovine lung endothelin receptor using biotinylated endothelin and avidin-agarose.
PMID 1325465
Abstract
Endothelin receptor was purified from bovine lung by affinity chromatography using biotinylated endothelin and avidin-agarose. Endothelin was biotinylated with sulphosuccinimidyl 2-(biotinamido)ethyl-1,3-dithiopropionate, a reactive form of biotin with a cleavable spacer arm containing a disulphide bond designed for a simple elution by thiols. Starting from 3.5 kg of bovine lung, about 200 micrograms of pure receptor were obtained.