EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Cytoskeletal integrity in interphase cells requires protein phosphatase activity.
PMID 1332069
Abstract
Phosphorylation by protein kinases has been established as a key factor in the regulation of cytoskeletal structure. However, little is known about the role of protein phosphatases in cytoskeletal regulation. To assess the possible functions of protein phosphatases in this respect, we studied the effects of the phosphatase inhibitors calyculin A, okadaic acid, and dinophysistoxin 1 (35-methylokadaic acid) on BHK-21 fibroblasts. Within minutes of incubation with these inhibitors, changes are seen in the structural organization of intermediate filaments, followed by a loss of microtubules, as assayed by immunofluorescence. These changes in cytoskeletal structure are accompanied by a rapid and selective increase in vimentin phosphorylation on interphase-specific sites, and they are fully reversible after removal of calyculin A. The results indicate that there is a rapid phosphate turnover on cytoskeletal intermediate filaments and further suggest that protein phosphatases are essential for the maintenance and structural integrity of two major cytoskeletal components.