EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Heteromeric NMDA receptors: molecular and functional distinction of subtypes.
PMID 1350383
Abstract
The N-methyl D-aspartate (NMDA) receptor subtype of glutamate-gated ion channels possesses high calcium permeability and unique voltage-dependent sensitivity to magnesium and is modulated by glycine. Molecular cloning identified three complementary DNA species of rat brain, encoding NMDA receptor subunits NMDAR2A (NR2A), NR2B, and NR2C, which are 55 to 70% identical in sequence. These are structurally related, with less than 20% sequence identity, to other excitatory amino acid receptor subunits, including the NMDA receptor subunit NMDAR1 (NR1). Upon expression in cultured cells, the new subunits yielded prominent, typical glutamate- and NMDA-activated currents only when they were in heteromeric configurations with NR1. NR1-NR2A and NR1-NR2C channels differed in gating behavior and magnesium sensitivity. Such heteromeric NMDA receptor subtypes may exist in neurons, since NR1 messenger RNA is synthesized throughout the mature rat brain, while NR2 messenger RNA show a differential distribution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB5046P
Anti-NMDAR1 Splice Variant C1 Antibody, Anti-NMDAR1 Splice Variant C1 Antibody is an antibody against NMDAR1 Splice Variant C1 for use in WB.
AB5048P
Anti-NMDAR1 Splice Variant C2 Antibody, This Anti-NMDAR1 Splice Variant C2 Antibody is validated for use in IH, WB for the detection of NMDAR1 Splice Variant C2.
AB5050P
Anti-NMDAR1 Splice Variant C2 Antibody, Detect NMDAR1 Splice Variant C2 using this Anti-NMDAR1 Splice Variant C2 Antibody validated for use in IH & WB.
G0166 Anti-Glutamate Receptor NMDAR1 (NR1) (Splice Variant Insert C1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
G0541 Anti-Glutamate Receptor NMDAR1 (NR1) (Splice Variant Insert N1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
G8913
Anti-Glutamate Receptor NMDAR1 (NR1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
G9038 Anti-Glutamate Receptor NMDAR2A (NR2A) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
M264 Anti-Glutamate Receptor NMDAR2A (NR2A) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
M265 Anti-Glutamate Receptor NMDAR2B (NR2B) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
M266 Anti-Glutamate Receptor NMDAR2C (NR2C) antibody produced in rabbit, affinity isolated antibody, lyophilized powder