EMAIL THIS PAGE TO A FRIEND
Bulletin of the World Health Organization
A preliminary report on a selective medium for the culture of Brucella, including fastidious types.
PMID 13585070
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3051 Brucella broth base, for microbiology