EMAIL THIS PAGE TO A FRIEND
Experimental cell research
The enzymatic composition of rat liver microsomes during liver regeneration.
PMID 13815291
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0567 Microsomes from Liver, Pooled, from human male