EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Cleavage of the haem-protein link by acid methylethylketone.
PMID 13837237
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6278
Peroxidase Inactivated, ≤0.02 units/mg protein (biuret)