EMAIL THIS PAGE TO A FRIEND
American journal of hygiene
Preparation of poliovirus vaccines in a human fetal diploid cell strain.
PMID 13905660
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90020107
WI 38 Cell Line human, caucasian fibroblast-like fetal lung cell