EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Metabolism of gamma-hydroxyglutamic acid. I. Conversion to alpha-hydroxy-gamma-ketoglutarate by purified glutamic-aspartic transaminase to rat liver.
PMID 13948827
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76157
(4R)-4-Hydroxy-L-glutamic acid, ≥98.0% (TLC)
C5H9NO5