EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of clinical and laboratory investigation
Determination of cardiac output with cardio green in a direct writing colorimeter.
PMID 13956020
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21980
Cardiogreen, for microscopy
C43H47N2NaO6S2