EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Metal ion complexing properties of carcinogen metabolites.
PMID 13999682
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
394114
2-Amino-4-phenylphenol, technical grade, 90%
C12H11NO