EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
STUDIES ON POLYNUCLEOTIDES. XXII. ENZYMIC DEGRADATION. AN EXONUCLEASE FROM LACTOBACILLUS ACIDOPHILUS R26. A. PURIFICATION, PROPERTIES, AND SUBSTRATE SPECIFICITY.
PMID 14063303