EMAIL THIS PAGE TO A FRIEND
Experimental cell research
VIRUSES AND MAMMALIAN CHROMOSOMES; CHROMOSOME ABERRATIONS BY HUMAN ADENOVIRUS TYPE 12.
PMID 14165598
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87091502
DEDE