EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
AN IMPROVED STAINING METHOD FOR ELECTRON MICROSCOPY.
PMID 14206430
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1775
Methanol, Absolute - Acetone free
CH4O