EMAIL THIS PAGE TO A FRIEND
The Anatomical record
A RELIABLE AND EASILY SECTIONED EPOXY EMBEDDING MEDIUM.
PMID 14223588
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3415
Bisphenol A diglycidyl ether
C21H24O4
31185
D.E.R. 332, used as embedding medium
C21H24O4