EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
INTERACTION BETWEEN DENATURED DNA, POLYRIBONUCLEOTIDES, AND RIBOSOMAL RNA: ATTEMPTS AT PREPARATIVE SEPARATION OF THE COMPLEMENTARY DNA STRANDS.
PMID 14229083