EMAIL THIS PAGE TO A FRIEND
Hoppe-Seyler's Zeitschrift fur physiologische Chemie
[ON THE DETERMINATION OF TRYPSIN AND CHYMOTRYPSIN WITH AMINO-P-NITROANILIDES].
PMID 14312556
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2628
N-Succinyl-L-phenylalanine-p-nitroanilide, protease substrate
C19H19N3O6