EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Biochemical changes in carbon tetrachloride fatty liver: concentration of carbon tetrachloride in liver and blood.
PMID 14436774
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02671
Carbon tetrachloride, analytical standard
CCl4