EMAIL THIS PAGE TO A FRIEND
Virology
Polyoma transformation of hamster cell clones--an investigation of genetic factors affecting cell competence.
PMID 14468055
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
84100501
BHK21C13-3P