EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Optimal conditions for the enzymatic determination of L-lactic acid.
PMID 14481932
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
440469
Lithium lactate, 95%
C3H5LiO3