EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
A simple method for the preparation of D-ribulose 5-phosphate.
PMID 14487826
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R9875
D-Ribulose 5-phosphate sodium salt, ≥90%
C5H11O8P · xNa+