EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis, in vitro evaluation, and intraocular pressure effects of water-soluble prodrugs of endocannabinoid noladin ether.
PMID 14584958
Abstract
The poor aqueous solubility of 2-arachidonyl glyceryl ether (noladin ether) 2 hinders both pharmacological studies and pharmaceutical development. The synthesized mono- and diphosphate esters of noladin ether (4 and 6) considerably increased the aqueous solubility of noladin ether (>40000-fold), showed high stability against chemical hydrolysis in buffer solutions, and were rapidly converted to the parent drug via enzymatic hydrolysis. The monophosphate ester of noladin ether reduced intraocular pressure in normotensive rabbits.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
437891
Dimethyl chlorophosphate, 96%
C2H6ClO3P