EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Palladium charcoal-catalyzed Suzuki-Miyaura coupling to obtain arylpyridines and arylquinolines.
PMID 14629166
Abstract
A phosphine ligand, such as PPh3 or 2-(dicyclohexylphosphino)biphenyl, is essential for the Pd/C-catalyzed Suzuki-Miyaura coupling of halopyridines and haloquinolines, although it has been reported that the reaction of phenyl chlorides can be catalyzed by nonprereduced Pd/C without any additives. In the reactions of bromopyridines, bromoquinolines, 2-chloropyridines, and 2-chloroquinolines, PPh3 was effective enough to provide coupling products in good yields. However, in the reactions of 3-chloropyridine, 4-chloropyridine, and 6-chloroquinoline, sterically hindered 2-(dicyclohexylphosphino)biphenyl was indispensable as a ligand.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
375098
6-Chloroquinoline, 99%
C9H6ClN