EMAIL THIS PAGE TO A FRIEND
FEBS letters
Potassium channels.
PMID 14630320
Abstract
The atomic structures of K+ channels have added a new dimension to our understanding of K+ channel function. I will briefly review how structures have influenced our views on ion conduction, gating of the pore, and voltage sensing.