EMAIL THIS PAGE TO A FRIEND
Genes and immunity
Cloning and characterization of GITR ligand.
PMID 14647196
Abstract
The gene encoding the natural ligand of murine glucocorticoid-induced tumor necrosis factor receptor (GITR) was cloned and characterized. The putative GITR ligand (GITRL) is composed of 173 amino acids with features resembling those of type II membrane proteins and is 51% identical to the human activation-inducible TNF receptor (AITR) ligand, TL6. Expression of the GITRL is restricted to immature and mature splenic dendritic cells. GITRL binds GITR expressed on HEK 293 cells and triggers NF-kappaB activation. Functional studies reveal that soluble CD8-GITRL prevents CD4+CD25+ regulatory T-cell-mediated suppressive activities.