EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
The second metal-binding site of 70 kDa heat-shock protein is essential for ADP binding, ATP hydrolysis and ATP synthesis.
PMID 14664695
Abstract
The chaperone activity of Hsp70 (70 kDa heat-shock protein) in protein folding and its conformational switch, including oligomeric and monomeric interconversion, are regulated by the hydrolysis of ATP and the ATP-ADP exchange cycle. The crystal structure of human ATPase domain shows two metal-binding sites, the first for ATP binding and a second, in close proximity to the first, whose function remains unknown [Sriram, Osipiuk, Freeman, Morimoto and Joachimiak (1997) Structure 5, 403-414]. In this study, we have characterized the second metal-binding motif by site-directed mutagenesis and the kinetics of ATP and ADP binding, and found that the second metal-binding site, comprising a loop co-ordinated by His-227, Glu-231 and Asp-232, participates both in ATP hydrolysis and ATP-synthetic activities, in co-operation with the first metal-binding site. The first metal-binding site, a catalytic centre, is essential for ATP binding and the second site for ADP binding in the reactions of ATP hydrolysis and ATP synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2810
Adenosine 5′-diphosphate–Agarose, lyophilized powder