EMAIL THIS PAGE TO A FRIEND
Oncogene
Overexpression of LRP12, a gene contained within an 8q22 amplicon identified by high-resolution array CGH analysis of oral squamous cell carcinomas.
PMID 14676824
Abstract
Chromosome 8q amplification is a common event observed in cancer. In this study, we used high-resolution array comparative genomic hybridization to resolve two neighboring regions on 8q that are both amplified in oral cancer. One region (at 8q24) contains the MYC oncogene, which is frequently overexpressed in many cancers, while the other region (at 8q22) represents a novel amplicon. The alignment of array comparative genomic hybridization profiles of 20 microdissected oral squamous cell carcinomas (OSCCs) revealed a approximately 5 Mbp region of frequent copy number alteration. This region harbors 16 known genes. Gene expression analysis comparing 15 microdissected OSCC with 16 normal epithelium samples revealed overexpression specific to LRP12 but not the neighboring genes, dihydropyrimidinase and FOG2, suggesting that LRP12 may function as an oncogene in oral tumors.