EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Silica-water reaction media: its application to the formation and ring opening of aziridines.
PMID 14694476
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
402869
Chloramine T trihydrate, ACS reagent, 98%
C7H7ClNNaO2S · 3H2O
857319
Chloramine-T hydrate, 95%
C7H7ClNNaO2S · xH2O