EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Linear free energy correlation analysis on the electronic effects of Rh(II) carbene O-H insertion.
PMID 14703404
Abstract
The relative rate constants for the Rh(II)-catalyzed insertion of diazoacetone into the O-H bond have been measured through intermolecular competitions. The kinetic data were subjected to Hammett correlation analysis, and mechanistic implication of the results with respect to a stepwise vs a concerted O-H insertion pathway is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209058
Rhodium(II) acetate dimer, powder
C8H12O8Rh2