EMAIL THIS PAGE TO A FRIEND
Genome research
An abundance of bidirectional promoters in the human genome.
PMID 14707170
Abstract
The alignment of full-length human cDNA sequences to the finished sequence of the human genome provides a unique opportunity to study the distribution of genes throughout the genome. By analyzing the distances between 23,752 genes, we identified a class of divergently transcribed gene pairs, representing more than 10% of the genes in the genome, whose transcription start sites are separated by less than 1000 base pairs. Although this bidirectional arrangement has been previously described in humans and other species, the prevalence of bidirectional gene pairs in the human genome is striking, and the mechanisms of regulation of all but a few bidirectional genes are unknown. Our work shows that the transcripts of many bidirectional pairs are coexpressed, but some are antiregulated. Further, we show that many of the promoter segments between two bidirectional genes initiate transcription in both directions and contain shared elements that regulate both genes. We also show that the bidirectional arrangement is often conserved among mouse orthologs. These findings demonstrate that a bidirectional arrangement provides a unique mechanism of regulation for a significant number of mammalian genes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HUTR001C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Housekeeping Control
HUTR002C
MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Empty Vector Control
HUTR003C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Non-target Control 1
HUTR004C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Non-target Control 2
HUTR00012 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ARNTL
HUTR00018 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RORA
HUTR00022 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, STAT3
HUTR00023 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TP53
HUTR00030 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TRIAP1
HUTR00036 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, MPP5
HUTR00041 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ARFIP1
HUTR00043 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, DHX40
HUTR00046 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, LATS2
HUTR00047 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RBL2
HUTR00238 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ATP5G2
HUTR00247 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, NDST3
HUTR00256 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, WNK2
HUTR00264 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, EDF1
HUTR00276 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, XCR1
HUTR00286 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RAB40A
HUTR00297 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TTR
HUTR00307 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, IFI27L1
HUTR00317 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, AMDHD2
HUTR00331 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, SHKBP1
HUTR00338 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CCDC123
HUTR00345 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CCT6B
HUTR00484 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TP73
HUTR00491 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TLR1
HUTR00499 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, SPATA5L1
HUTR00514 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CGB1